Product Details

SNP ID
rs60100877
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:44349558 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGCTGCCCCTTCCCAGAGGTGAGC[A/G]TGCCATCAGCCCAGTGGAGGGGCTT
Phenotype
MIM: 138945
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM171A2 PubMed Links

Gene Details

Gene
FAM171A2
Gene Name
family with sequence similarity 171 member A2
There are no transcripts associated with this gene.

Gene
GRN
Gene Name
granulin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002087.3 Intron NP_002078.1
XM_005257253.1 Intron XP_005257310.1

View Full Product Details