Product Details
- SNP ID
-
hCV89395975
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:30991695 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AACTGGCTTCTGGATACAGCAGGAA[A/T]TTTCAGCAACTAGGCTTACAGAGAA
- Phenotype
-
MIM: 600271
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
DSC3
PubMed Links
Gene Details
- Gene
- DSC3
- Gene Name
- desmocollin 3
View Full Product Details