Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001127174.1 | 1802 | UTR 3 | NP_001120646.1 | ||
NM_001127175.1 | 1802 | UTR 3 | NP_001120647.1 | ||
NM_001127176.1 | 1802 | UTR 3 | NP_001120648.1 | ||
NM_031939.3 | 1802 | UTR 3 | NP_114145.2 | ||
XM_005258365.3 | 1802 | UTR 3 | XP_005258422.1 | ||
XM_011526226.2 | 1802 | UTR 3 | XP_011524528.1 | ||
XM_017026035.1 | 1802 | UTR 3 | XP_016881524.1 | ||
XM_017026036.1 | 1802 | UTR 3 | XP_016881525.1 | ||
XM_017026037.1 | 1802 | UTR 3 | XP_016881526.1 |