Product Details

SNP ID
rs58761658
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:37508330 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCATCCCAAGAGTGAGGCTGGTTTC[C/T]GGAAATCCATTCTGACTCTGTGACA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF793 PubMed Links

Gene Details

Gene
ZNF793
Gene Name
zinc finger protein 793
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013659.2 245 UTR 5 NP_001013681.2
XM_005258927.3 245 UTR 5 XP_005258984.1
XM_006723213.3 245 UTR 5 XP_006723276.1
Gene
ZNF793-AS1
Gene Name
ZNF793 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details