Product Details

SNP ID
rs61353573
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:37522627 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCAGTGTTCATCTTCCATGAAGT[C/T]GACATTTTTGAAGAGTATAGGTAGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF793 PubMed Links

Gene Details

Gene
ZNF793
Gene Name
zinc finger protein 793
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013659.2 414 UTR 5 NP_001013681.2
XM_005258927.3 414 UTR 5 XP_005258984.1
XM_006723213.3 414 UTR 5 XP_006723276.1

View Full Product Details