Product Details

SNP ID
rs62111265
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:45890929 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGGGAGTCAACTGGGGTTCCTGG[C/T]AGAAGGGGCAGCAGAGCGCTGAGGG
Phenotype
MIM: 615331
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IRF2BP1 PubMed Links

Gene Details

Gene
IRF2BP1
Gene Name
interferon regulatory factor 2 binding protein 1
There are no transcripts associated with this gene.

Gene
MYPOP
Gene Name
Myb related transcription factor, partner of profilin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012643.3 981 Silent Mutation NP_001012661.1
XM_017026742.1 981 Silent Mutation XP_016882231.1
XM_017026743.1 981 UTR 3 XP_016882232.1

View Full Product Details