Product Details

SNP ID
rs62128691
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:47527311 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACTTAGCAAATCTTATTATACTGC[A/G]ATTATGTGCAATTTTGTATACATCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF541 PubMed Links

Gene Details

Gene
ZNF541
Gene Name
zinc finger protein 541
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277075.1 Intron NP_001264004.1
XM_005259311.4 Intron XP_005259368.1
XM_005259314.3 Intron XP_005259371.1
XM_011527367.2 Intron XP_011525669.1
XM_011527368.2 Intron XP_011525670.1
XM_011527369.2 Intron XP_011525671.1
XM_011527370.2 Intron XP_011525672.1
XM_011527371.2 Intron XP_011525673.1
XM_011527372.2 Intron XP_011525674.1
XM_011527373.2 Intron XP_011525675.1
XM_011527374.2 Intron XP_011525676.1
XM_011527375.1 Intron XP_011525677.1
XM_017027351.1 Intron XP_016882840.1

View Full Product Details