Product Details

SNP ID
rs59356506
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:49659853 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACAACCTGCAGGGGAAGTGGGGAC[A/G]GGAGTCAGGGAAAACACCCAGCCAC
Phenotype
MIM: 610837 MIM: 603734
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BCL2L12 PubMed Links

Gene Details

Gene
BCL2L12
Gene Name
BCL2 like 12
There are no transcripts associated with this gene.

Gene
IRF3
Gene Name
interferon regulatory factor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001197122.1 Intron NP_001184051.1
NM_001197123.1 Intron NP_001184052.1
NM_001197124.1 Intron NP_001184053.1
NM_001197125.1 Intron NP_001184054.1
NM_001197126.1 Intron NP_001184055.1
NM_001197127.1 Intron NP_001184056.1
NM_001197128.1 Intron NP_001184057.1
NM_001571.5 Intron NP_001562.1
XM_006723197.1 Intron XP_006723260.1
XM_006723198.1 Intron XP_006723261.1
XM_006723200.1 Intron XP_006723263.1
XM_006723201.1 Intron XP_006723264.1
XM_006723202.2 Intron XP_006723265.1
XM_017026766.1 Intron XP_016882255.1
XM_017026767.1 Intron XP_016882256.1
Gene
SCAF1
Gene Name
SR-related CTD associated factor 1
There are no transcripts associated with this gene.

View Full Product Details