Product Details
- SNP ID
-
rs62125391
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:2904245 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTTTTCTGTGGTTTCTTTCCTTTGC[A/G]CACATGCTGGTGTCTGAGACAGTGG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF57
PubMed Links
Gene Details
- Gene
- ZNF57
- Gene Name
- zinc finger protein 57
View Full Product Details