Product Details

SNP ID
rs56801248
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:3478420 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCAGCATCCGCACGCATAGTACC[C/T]CTGGTCCTGGTATAGGTTGGACTCC
Phenotype
MIM: 600729
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NFIC PubMed Links

Gene Details

Gene
NFIC
Gene Name
nuclear factor I C
There are no transcripts associated with this gene.

Gene
SMIM24
Gene Name
small integral membrane protein 24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136503.1 317 Missense Mutation AGT,GGT S80G NP_001129975.1

View Full Product Details