Product Details

SNP ID
rs62131833
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:10858673 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCTTCCCCACACCCCTTTACCTC[C/T]AGCTGCCAGAAAACGGTCACAATCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C19orf38 PubMed Links

Gene Details

Gene
C19orf38
Gene Name
chromosome 19 open reading frame 38
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136482.1 Intron NP_001129954.1
XM_005259846.4 Intron XP_005259903.1
XM_005259847.4 Intron XP_005259904.1

View Full Product Details