Product Details
- SNP ID
-
rs57208996
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:15453304 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCGCAGATTGGCGGCGGCGGGCAG[G/T]ACGCCGGACCGGGACACTCTGCGTG
- Phenotype
-
MIM: 616561
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
MIR1470
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs138850805] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MIR1470
- Gene Name
- microRNA 1470
There are no transcripts associated with this gene.
- Gene
- RASAL3
- Gene Name
- RAS protein activator like 3
- Gene
- WIZ
- Gene Name
- widely interspaced zinc finger motifs
There are no transcripts associated with this gene.
View Full Product Details