Product Details

SNP ID
rs61760056
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:63696958 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTTTGTGCAGCGGCCGTGCCGCC[A/G]AGACAGCCCCACGACGTGTGGCCCG
Phenotype
MIM: 604699 MIM: 608833 MIM: 603361
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARFRP1 PubMed Links

Gene Details

Gene
ARFRP1
Gene Name
ADP ribosylation factor related protein 1
There are no transcripts associated with this gene.

Gene
RTEL1
Gene Name
regulator of telomere elongation helicase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001283009.1 308 Intron NP_001269938.1
NM_001283010.1 308 Intron NP_001269939.1
NM_016434.3 308 Intron NP_057518.1
NM_032957.4 308 Intron NP_116575.3
Gene
RTEL1-TNFRSF6B
Gene Name
RTEL1-TNFRSF6B readthrough (NMD candidate)
There are no transcripts associated with this gene.

Gene
TNFRSF6B
Gene Name
TNF receptor superfamily member 6b
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003823.3 308 Missense Mutation CAA,CGA Q64R NP_003814.1

View Full Product Details