Product Details

SNP ID
rs56145017
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:63529586 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCTGGAGAGCCTCTCCCGCAGGG[C/T]CTTGAAGCAGGGTCTCTGCTCGGGG
Phenotype
MIM: 602004
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PPDPF PubMed Links

Gene Details

Gene
PPDPF
Gene Name
pancreatic progenitor cell differentiation and proliferation factor
There are no transcripts associated with this gene.

Gene
PTK6
Gene Name
protein tyrosine kinase 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256358.1 1240 UTR 3 NP_001243287.1
NM_005975.3 1240 Missense Mutation ACC,GCC T436A NP_005966.1
XM_017027982.1 1240 Intron XP_016883471.1

View Full Product Details