Product Details

SNP ID
rs60374965
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:62390024 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCAAGTGTGTTTTCAAGTCAACTT[C/T]AGCAGACCTCATTTAACCATTTTTT
Phenotype
MIM: 180477
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CABLES2 PubMed Links

Gene Details

Gene
CABLES2
Gene Name
Cdk5 and Abl enzyme substrate 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031215.2 2391 UTR 3 NP_112492.2
Gene
RPS21
Gene Name
ribosomal protein S21
There are no transcripts associated with this gene.

View Full Product Details