Product Details

SNP ID
rs62223962
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:28929413 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCAGGTTTTCACTTACTCCACTT[C/T]AGCCTATTATACTGAATTTTACCTA
Phenotype
MIM: 613083
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LTN1 PubMed Links

Gene Details

Gene
LTN1
Gene Name
listerin E3 ubiquitin protein ligase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320766.1 6448 UTR 3 NP_001307695.1
NM_015565.2 6448 UTR 3 NP_056380.2
XM_006723987.3 6448 Intron XP_006724050.1
XM_017028316.1 6448 UTR 3 XP_016883805.1
XM_017028317.1 6448 UTR 3 XP_016883806.1

View Full Product Details