Product Details

SNP ID
rs62229399
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.21:36337892 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCCTTTTGTCTCAGACCTACTTG[A/G]AAGTCATAAAAGCGGAGCATGTTGT
Phenotype
MIM: 610078
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
MORC3 PubMed Links

Gene Details

Gene
MORC3
Gene Name
MORC family CW-type zinc finger 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320445.1 409 Missense Mutation AAA,GAA K65E NP_001307374.1
NM_001320446.1 409 Missense Mutation AAA,GAA K65E NP_001307375.1
NM_015358.2 409 Missense Mutation AAA,GAA K136E NP_056173.1
XM_011529518.2 409 Missense Mutation AAA,GAA K65E XP_011527820.1
XM_017028311.1 409 Missense Mutation AAA,GAA K65E XP_016883800.1
XM_017028312.1 409 UTR 5 XP_016883801.1

View Full Product Details