Product Details

SNP ID
rs55742502
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:45859847 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTCAGCTGAGGAAGCAGGGCAGG[C/T]GGCCTCTTCCTTCAACACCTTTAAC
Phenotype
MIM: 608502
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PCBP3 PubMed Links

Gene Details

Gene
PCBP3
Gene Name
poly(rC) binding protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130141.1 Intron NP_001123613.1
NM_020528.2 Intron NP_065389.2

View Full Product Details