Product Details

SNP ID
rs59991531
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:39532705 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTCACCCTCGGACGCCGCCAGCA[G/T]CTCCAGGCCCCGCCGCAGCAGGGCG
Phenotype
MIM: 610225
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
RPS19BP1 PubMed Links

Gene Details

Gene
RPS19BP1
Gene Name
ribosomal protein S19 binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_194326.3 151 Missense Mutation NP_919307.1

View Full Product Details