Product Details

SNP ID
rs58176776
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:103679860 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGTTTGTTAGTAGCACCTGTGGG[A/C]AAAAAAAAAAAAAAAAAAAAAAAAA
Phenotype
MIM: 300409
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MORF4L2 PubMed Links

Gene Details

Gene
MORF4L2
Gene Name
mortality factor 4 like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142418.1 Intron NP_001135890.1
NM_001142419.1 Intron NP_001135891.1
NM_001142420.1 Intron NP_001135892.1
NM_001142421.1 Intron NP_001135893.1
NM_001142422.1 Intron NP_001135894.1
NM_001142423.1 Intron NP_001135895.1
NM_001142424.1 Intron NP_001135896.1
NM_001142425.1 Intron NP_001135897.1
NM_001142426.1 Intron NP_001135898.1
NM_001142427.1 Intron NP_001135899.1
NM_001142428.1 Intron NP_001135900.1
NM_001142429.1 Intron NP_001135901.1
NM_001142430.1 Intron NP_001135902.1
NM_001142431.1 Intron NP_001135903.1
NM_001142432.1 Intron NP_001135904.1
NM_012286.2 Intron NP_036418.1
Gene
MORF4L2-AS1
Gene Name
MORF4L2 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details