Product Details
- SNP ID
-
rs62612880
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:83866524 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGTCCGTTTAATTCTTTCATCCTTC[A/C]TCTCTCTGTCCATTTGCATCCCAAA
- Phenotype
-
MIM: 300768
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CYLC1
PubMed Links
Gene Details
- Gene
- CYLC1
- Gene Name
- cylicin 1
View Full Product Details