Product Details

SNP ID
rs55962594
Assay Type
Functionally Tested
NCBI dbSNP Submissions
5
Location
Chr.1:207506782 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCATGTGATCAAAGGCATCCAGTTC[A/G]GATCCCAAATTAAATATTCTTGTAC
Phenotype
MIM: 120620
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
CR1 PubMed Links

Gene Details

Gene
CR1
Gene Name
complement component 3b/4b receptor 1 (Knops blood group)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000573.3 486 Missense Mutation AGA,GGA R124G NP_000564.2
NM_000651.4 486 Missense Mutation AGA,GGA R124G NP_000642.3
XM_006711166.2 486 Missense Mutation AGA,GGA R129G XP_006711229.1
XM_011509205.1 486 Missense Mutation AGA,GGA R129G XP_011507507.1

View Full Product Details