Product Details
- SNP ID
-
rs61821322
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
1
- Location
-
Chr.1:207887658 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGTTGGGCGTAAGAGATGTCACCTC[A/C]AGCATGGGGGTAGCACGTGGTCAGA
- Phenotype
-
MIM: 142230
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CD34
PubMed Links
Gene Details
- Gene
- CD34
- Gene Name
- CD34 molecule
View Full Product Details