Product Details

SNP ID
rs57765111
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:6524410 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTTAAATGCAGAAATGATGTGAC[C/T]TGGTCCCATCACCCGTTTACACAAA
Phenotype
MIM: 611101
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NOL9 PubMed Links

Gene Details

Gene
NOL9
Gene Name
nucleolar protein 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024654.4 3649 UTR 3 NP_078930.3
XM_005263493.3 3649 Intron XP_005263550.1
XM_011542147.2 3649 Intron XP_011540449.1
XM_017002337.1 3649 Intron XP_016857826.1
XM_017002338.1 3649 Intron XP_016857827.1
XM_017002339.1 3649 Intron XP_016857828.1
Gene
PLEKHG5
Gene Name
pleckstrin homology and RhoGEF domain containing G5
There are no transcripts associated with this gene.

View Full Product Details