Product Details

SNP ID
rs56761572
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:86815743 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTGCAAAAAGAAAAACAAGAGAG[A/T]CTCAATACATGCACCAAGTCAAGGT
Phenotype
MIM: 186910 MIM: 186730
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CD8A PubMed Links

Gene Details

Gene
CD8A
Gene Name
CD8a molecule
There are no transcripts associated with this gene.

Gene
CD8B
Gene Name
CD8b molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001178100.1 Intron NP_001171571.1
NM_004931.4 Intron NP_004922.1
NM_172101.3 Intron NP_742099.1
NM_172102.3 Intron NP_742100.1
NM_172213.3 Intron NP_757362.1
XM_011533164.2 Intron XP_011531466.1

View Full Product Details