Product Details

SNP ID
rs56204436
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:24117532 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTGGGCCAGTGAGAATTAGGCCA[A/G]AATTTTCATTAGAATTGTTGGAGAA
Phenotype
MIM: 190160
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
THRB PubMed Links

Gene Details

Gene
THRB
Gene Name
thyroid hormone receptor beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000461.4 7160 UTR 3 NP_000452.2
NM_001128176.2 7160 UTR 3 NP_001121648.1
NM_001128177.1 7160 UTR 3 NP_001121649.1
NM_001252634.1 7160 UTR 3 NP_001239563.1
XM_005265421.4 7160 Intron XP_005265478.1
XM_005265423.4 7160 Intron XP_005265480.1
XM_005265424.3 7160 Intron XP_005265481.1
XM_006713317.3 7160 Intron XP_006713380.1
XM_006713318.3 7160 Intron XP_006713381.1
XM_011534046.2 7160 Intron XP_011532348.1
XM_011534047.2 7160 Intron XP_011532349.1
XM_011534048.2 7160 Intron XP_011532350.1
XM_011534049.2 7160 Intron XP_011532351.1
XM_011534050.2 7160 Intron XP_011532352.1
XM_011534051.2 7160 Intron XP_011532353.1
XM_011534052.2 7160 Intron XP_011532354.1
XM_011534053.2 7160 Intron XP_011532355.1
XM_017007108.1 7160 Intron XP_016862597.1
XM_017007109.1 7160 Intron XP_016862598.1
XM_017007110.1 7160 Intron XP_016862599.1
XM_017007111.1 7160 Intron XP_016862600.1
XM_017007112.1 7160 Intron XP_016862601.1
XM_017007113.1 7160 Intron XP_016862602.1
XM_017007114.1 7160 Intron XP_016862603.1
XM_017007115.1 7160 Intron XP_016862604.1

View Full Product Details