Product Details

SNP ID
rs62245251
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:36831028 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCGATGTCCTGCACCACCAGCTTG[A/C]CTTCATCAATGGCCGGGTCCACCTT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TRANK1 PubMed Links

Gene Details

Gene
TRANK1
Gene Name
tetratricopeptide repeat and ankyrin repeat containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014831.2 9600 Missense Mutation GGC,GTC G2808V NP_055646.2
XM_006713432.3 9600 Missense Mutation GGC,GTC G2852V XP_006713495.1
XM_011534289.2 9600 Missense Mutation GGC,GTC G2853V XP_011532591.1
XM_017007570.1 9600 Missense Mutation GGC,GTC G2819V XP_016863059.1
XM_017007571.1 9600 Missense Mutation GGC,GTC G2876V XP_016863060.1
XM_017007572.1 9600 Missense Mutation GGC,GTC G2844V XP_016863061.1
XM_017007573.1 9600 Intron XP_016863062.1

View Full Product Details