Product Details

SNP ID
rs55884193
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:52221415 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCGTCAGGGCTCAGGATCACCAG[C/G]ACCACGACGTCCTTGCGGTCCTCCA
Phenotype
MIM: 125290 MIM: 605474 MIM: 607433
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ALAS1 PubMed Links

Gene Details

Gene
ALAS1
Gene Name
5'-aminolevulinate synthase 1
There are no transcripts associated with this gene.

Gene
TLR9
Gene Name
toll like receptor 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017442.3 3535 Silent Mutation GTC,GTG V967V NP_059138.1
Gene
TWF2
Gene Name
twinfilin actin binding protein 2
There are no transcripts associated with this gene.

View Full Product Details