Product Details

SNP ID
rs61017521
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:57630920 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACCCAAATAGATTTGAATATATT[A/C]ATACCTTCCATGGGGAAATACTTTT
Phenotype
MIM: 616519
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DENND6A PubMed Links

Gene Details

Gene
DENND6A
Gene Name
DENN domain containing 6A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152678.2 Intron NP_689891.1
XM_006713019.3 Intron XP_006713082.1
XM_006713020.2 Intron XP_006713083.1
XM_017005864.1 Intron XP_016861353.1
Gene
PDE12
Gene Name
phosphodiesterase 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322176.1 Intron NP_001309105.1
NM_001322177.1 Intron NP_001309106.1
NM_177966.6 Intron NP_808881.3

View Full Product Details