Product Details

SNP ID
rs55695573
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:8877609 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAAAAACAAAATCCGTCCATTGCA[C/T]GTTTGATAGTGCCATGAACCTCTCT
Phenotype
MIM: 605256
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RAD18 PubMed Links

Gene Details

Gene
RAD18
Gene Name
RAD18, E3 ubiquitin protein ligase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020165.3 5326 UTR 3 NP_064550.3
XM_017006873.1 5326 Intron XP_016862362.1

View Full Product Details