Product Details

SNP ID
rs59092768
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:175808877 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATCAAGAGCAGCAGTGGTGGCCTC[C/T]GGAGAAGTGAGGGCATGCCTTCACT
Phenotype
MIM: 605033
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CPLX2 PubMed Links

Gene Details

Gene
CPLX2
Gene Name
complexin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008220.1 Intron NP_001008221.1
NM_006650.3 Intron NP_006641.1
XM_005265798.1 Intron XP_005265855.1
XM_005265799.1 Intron XP_005265856.1
XM_011534419.1 Intron XP_011532721.1
XM_017008964.1 Intron XP_016864453.1

View Full Product Details