Product Details
- SNP ID
-
rs55910090
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:23682694 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCGTTCTGCGCCCCGGCGCGGCTGC[G/T]TCCTCCCTCTGGCTCTGGCTCTGGC
- Phenotype
-
MIM: 602041
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
NKX3-1
PubMed Links
Gene Details
- Gene
- NKX3-1
- Gene Name
- NK3 homeobox 1
View Full Product Details