Product Details

SNP ID
rs57111114
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:23682841 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTTGGACGGGGTCGGCGGCGCGG[C/T]CCCCTCCGCTTTCGCCTCCCCGGGC
Phenotype
MIM: 602041
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NKX3-1 PubMed Links

Gene Details

Gene
NKX3-1
Gene Name
NK3 homeobox 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256339.1 97 Intron NP_001243268.1
NM_006167.3 97 Missense Mutation ACC,GCC T17A NP_006158.2

View Full Product Details