Product Details

SNP ID
rs55754159
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:136673544 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCGAGTCCCTGCCCTCGAGCCCGG[A/G]GAGGGTCCAGCTGAGCCCCCTGCAG
Phenotype
MIM: 603100 MIM: 608582 MIM: 611767
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AGPAT2 PubMed Links

Gene Details

Gene
AGPAT2
Gene Name
1-acylglycerol-3-phosphate O-acyltransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012727.1 1051 UTR 3 NP_001012745.1
NM_006412.3 1051 UTR 3 NP_006403.2
Gene
EGFL7
Gene Name
EGF like domain multiple 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016215.4 1051 Intron NP_057299.1
NM_201446.2 1051 Intron NP_958854.1
XM_006717141.3 1051 Intron XP_006717204.1
XM_011518764.1 1051 Intron XP_011517066.1
XM_011518765.1 1051 Intron XP_011517067.1
XM_011518766.1 1051 Intron XP_011517068.1
XM_011518767.1 1051 Intron XP_011517069.1
XM_011518768.1 1051 Intron XP_011517070.1
XM_017014795.1 1051 Intron XP_016870284.1
XM_017014796.1 1051 Intron XP_016870285.1
XM_017014797.1 1051 Intron XP_016870286.1
XM_017014798.1 1051 Intron XP_016870287.1
XM_017014799.1 1051 Intron XP_016870288.1
Gene
MIR126
Gene Name
microRNA 126
There are no transcripts associated with this gene.

View Full Product Details