Product Details

SNP ID
rs61052533
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:44992787 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCTAGTTTTTTTTCTTGTTTATTC[C/T]GGTAAAATATACACATCTGTCATAT
Phenotype
MIM: 604354
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GPALPP1 PubMed Links

Gene Details

Gene
GPALPP1
Gene Name
GPALPP motifs containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001316951.1 Intron NP_001303880.1
NM_001316952.1 Intron NP_001303881.1
NM_018559.3 Intron NP_061029.2
XM_005266444.3 Intron XP_005266501.1
XM_011535144.2 Intron XP_011533446.1
Gene
NUFIP1
Gene Name
NUFIP1, FMR1 interacting protein 1
There are no transcripts associated with this gene.

View Full Product Details