Product Details

SNP ID
rs61961265
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:49989507 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCACTGATATTGTATAATTAGTTT[C/T]AACCATCTTCTGAATTATCTGTGTC
Phenotype
MIM: 605766 MIM: 605661
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DLEU2 PubMed Links

Gene Details

Gene
DLEU2
Gene Name
deleted in lymphocytic leukemia 2 (non-protein coding)
There are no transcripts associated with this gene.

Gene
MIR3613
Gene Name
microRNA 3613
There are no transcripts associated with this gene.

Gene
TRIM13
Gene Name
tripartite motif containing 13
There are no transcripts associated with this gene.

View Full Product Details