Product Details

SNP ID
rs60493263
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:25169058 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAATACCTGGCTCTTTCAGCTTTG[G/T]AGGTATTTTCCACACAGTGATACAG
Phenotype
MIM: 614659
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
AMER2 PubMed Links

Gene Details

Gene
AMER2
Gene Name
APC membrane recruitment protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152704.3 3116 UTR 3 NP_689917.2
NM_199138.2 3116 UTR 3 NP_954589.1
XM_005266279.3 3116 UTR 3 XP_005266336.1
XM_005266280.3 3116 UTR 3 XP_005266337.1
XM_017020415.1 3116 UTR 3 XP_016875904.1
Gene
LINC00463
Gene Name
long intergenic non-protein coding RNA 463
There are no transcripts associated with this gene.

View Full Product Details