Product Details

SNP ID
rs61944045
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:27982747 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTCCTTCCTCCATCCTGTCTTCT[A/C]AACTCTAAGTCTTCCTCAACTCGGA
Phenotype
MIM: 615804
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
URAD PubMed Links

Gene Details

Gene
URAD
Gene Name
ureidoimidazoline (2-oxo-4-hydroxy-4-carboxy-5-) decarboxylase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105577.1 Intron NP_001099047.1

View Full Product Details