Product Details

SNP ID
rs62002011
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:25116801 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGAAACATGTCCTTCACCAGTGGA[A/G]TTCTTAGAAGCTTTCCTTGGGCTTC
Phenotype
MIM: 601491
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IPW PubMed Links

Gene Details

Gene
IPW
Gene Name
imprinted in Prader-Willi syndrome (non-protein coding)
There are no transcripts associated with this gene.

Gene
SNORD116-30
Gene Name
small nucleolar RNA, C/D box 116-30
There are no transcripts associated with this gene.

View Full Product Details