Product Details

SNP ID
rs59633008
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:150221319 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAAGAGGAAAAGAAAGAGAGAATG[C/T]GAACCCGAGGCTGCAGGATGAGGCA
Phenotype
MIM: 114078
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CAMK2A PubMed Links

Gene Details

Gene
CAMK2A
Gene Name
calcium/calmodulin dependent protein kinase II alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015981.3 3043 UTR 3 NP_057065.2
NM_171825.2 3043 UTR 3 NP_741960.1
XM_017009898.1 3043 Intron XP_016865387.1

View Full Product Details