Product Details

SNP ID
rs62377911
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:151746678 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGTATTTTTTTTATAACTCTATT[C/T]CTGAACATACAAAAACAGGCTGTGA
Phenotype
MIM: 602270
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATOX1 PubMed Links

Gene Details

Gene
ATOX1
Gene Name
antioxidant 1 copper chaperone
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004045.3 Intron NP_004036.1

View Full Product Details