Product Details

SNP ID
rs55769974
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:48653888 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTGGGAAGAAACCCACGGTCCAC[A/G]TCCTCAAAATTTGGCTTTACGTTTG
Phenotype
MIM: 615488 MIM: 615487
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KANSL2 PubMed Links

Gene Details

Gene
KANSL2
Gene Name
KAT8 regulatory NSL complex subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017822.3 1710 UTR 3 NP_060292.3
Gene
MIR1291
Gene Name
microRNA 1291
There are no transcripts associated with this gene.

Gene
SNORA2A
Gene Name
small nucleolar RNA, H/ACA box 2A
There are no transcripts associated with this gene.

Gene
SNORA2C
Gene Name
small nucleolar RNA, H/ACA box 2C
There are no transcripts associated with this gene.

View Full Product Details