Product Details

SNP ID
rs61758378
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:48979627 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGCGTGAGTGGGGGGCTGCAGAG[A/T]GCCGTGCGCGAGTGCAAGTGGCAGT
Phenotype
MIM: 164820 MIM: 601906
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
WNT1 PubMed Links

Gene Details

Gene
WNT1
Gene Name
Wnt family member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005430.3 462 Missense Mutation AGA,AGT R88S NP_005421.1
Gene
WNT10B
Gene Name
Wnt family member 10B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003394.3 462 Intron NP_003385.2

View Full Product Details