Product Details

SNP ID
rs61922633
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:65050830 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACAGTGTAATTTTTTTTTTTTTA[A/T]TACTGAATCTCTGGAATAATGGTAA
Phenotype
MIM: 605186
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
WIF1 PubMed Links

Gene Details

Gene
WIF1
Gene Name
WNT inhibitory factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007191.4 2034 UTR 3 NP_009122.2

View Full Product Details