Product Details

SNP ID
rs55923552
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:195746915 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGTGTGTGTGTGTGTGCACGCGCG[C/T]GTGCACAGGCTAGTGTCCTTCTGTG
Phenotype
MIM: 158372
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MUC4 PubMed Links

Gene Details

Gene
MUC4
Gene Name
mucin 4, cell surface associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004532.5 3948 UTR 3 NP_004523.3
NM_018406.6 3948 Intron NP_060876.5
NM_138297.4 3948 UTR 3 NP_612154.2

View Full Product Details