Product Details

SNP ID
hCV90384308
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:110295377 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTCACGCTGCTGGGCAATCATTT[C/T]TTTTCTGCATGAAAAAAAGTTAAAT
Phenotype
MIM: 600020 MIM: 603519
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MXI1 PubMed Links

Gene Details

Gene
MXI1
Gene Name
MAX interactor 1, dimerization protein
There are no transcripts associated with this gene.

Gene
SMNDC1
Gene Name
survival motor neuron domain containing 1
There are no transcripts associated with this gene.

View Full Product Details