Product Details

SNP ID
rs60504087
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:13083422 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGTACAAAATACTGAAAGCCGCT[A/G]CAGGGGATTATAAAGATGTGTAAGA
Phenotype
MIM: 604258
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DLC1 PubMed Links

Gene Details

Gene
DLC1
Gene Name
DLC1 Rho GTPase activating protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164271.1 5727 UTR 3 NP_001157743.1
NM_001316668.1 5727 UTR 3 NP_001303597.1
NM_006094.4 5727 UTR 3 NP_006085.2
NM_024767.3 5727 Intron NP_079043.3
NM_182643.2 5727 UTR 3 NP_872584.2
XM_005273374.1 5727 UTR 3 XP_005273431.1
XM_017012951.1 5727 UTR 3 XP_016868440.1
XM_017012952.1 5727 UTR 3 XP_016868441.1

View Full Product Details