Product Details

SNP ID
rs58508726
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:42177573 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTGATCCTTCTAAGAGGCGCAAG[C/T]TGGCTGCCTTTCCAGAATCCTCTGA
Phenotype
MIM: 616035
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FOXJ3 PubMed Links

Gene Details

Gene
FOXJ3
Gene Name
forkhead box J3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198850.1 4222 UTR 3 NP_001185779.1
NM_001198851.1 4222 UTR 3 NP_001185780.1
NM_001198852.1 4222 UTR 3 NP_001185781.1
NM_014947.4 4222 UTR 3 NP_055762.3
XM_005270632.4 4222 UTR 3 XP_005270689.1
XM_006710458.3 4222 UTR 3 XP_006710521.1
XM_006710459.3 4222 UTR 3 XP_006710522.1
XM_011541026.2 4222 UTR 3 XP_011539328.1
XM_017000693.1 4222 UTR 3 XP_016856182.1
XM_017000694.1 4222 UTR 3 XP_016856183.1
XM_017000695.1 4222 UTR 3 XP_016856184.1

View Full Product Details