Product Details

SNP ID
rs58131089
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:43451806 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACCCCCTTCCACTTACCATTTGTA[A/T]TTGGAGGTTGGGTCTTCCTACCTTC
Phenotype
MIM: 615463
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
HYI PubMed Links

Gene Details

Gene
HYI
Gene Name
hydroxypyruvate isomerase (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190880.2 1070 Missense Mutation ATA,TTA I183L NP_001177809.1
NM_001243526.1 1070 Missense Mutation ATA,TTA I208L NP_001230455.1
NM_031207.5 1070 Missense Mutation ATA,TTA I183L NP_112484.3
XM_005271239.4 1070 Missense Mutation ATA,TTA I208L XP_005271296.1
XM_005271240.4 1070 Missense Mutation ATA,TTA I174L XP_005271297.1
XM_006710937.3 1070 Missense Mutation ATA,TTA I208L XP_006711000.1
XM_011542222.2 1070 Missense Mutation ATA,TTA I174L XP_011540524.1
Gene
MIR6735
Gene Name
microRNA 6735
There are no transcripts associated with this gene.

Gene
SZT2
Gene Name
seizure threshold 2 homolog (mouse)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015284.3 1070 UTR 3 NP_056099.3
XM_005270686.3 1070 Intron XP_005270743.1
XM_006710501.3 1070 Intron XP_006710564.1
XM_011541106.2 1070 Intron XP_011539408.1
XM_011541107.2 1070 Intron XP_011539409.1
XM_017000819.1 1070 Intron XP_016856308.1
XM_017000820.1 1070 Intron XP_016856309.1
XM_017000821.1 1070 Intron XP_016856310.1

View Full Product Details