Product Details

SNP ID
rs59136674
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:52896451 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGTGAAATGATGAAGGCAATCTGG[C/T]CACAAATCTTCCTTCTGGATCCTGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ECHDC2 PubMed Links

Gene Details

Gene
ECHDC2
Gene Name
enoyl-CoA hydratase domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198961.1 4944 UTR 3 NP_001185890.1
NM_001198962.1 4944 UTR 3 NP_001185891.1
NM_001319958.1 4944 UTR 3 NP_001306887.1
NM_018281.3 4944 UTR 3 NP_060751.2
XM_011541709.2 4944 UTR 3 XP_011540011.1
XM_011541710.2 4944 UTR 3 XP_011540012.1
XM_011541711.2 4944 UTR 3 XP_011540013.1
XM_011541712.2 4944 UTR 3 XP_011540014.1
XM_011541713.2 4944 UTR 3 XP_011540015.1
XM_011541715.2 4944 UTR 3 XP_011540017.1
XM_011541719.2 4944 UTR 3 XP_011540021.1
XM_011541720.2 4944 UTR 3 XP_011540022.1
XM_011541721.2 4944 UTR 3 XP_011540023.1
XM_011541722.2 4944 UTR 3 XP_011540024.1
XM_011541723.2 4944 UTR 3 XP_011540025.1
XM_011541724.2 4944 UTR 3 XP_011540026.1
XM_011541725.2 4944 UTR 3 XP_011540027.1
XM_011541726.2 4944 UTR 3 XP_011540028.1
XM_011541727.2 4944 UTR 3 XP_011540029.1
XM_017001638.1 4944 UTR 3 XP_016857127.1
XM_017001639.1 4944 UTR 3 XP_016857128.1
XM_017001640.1 4944 UTR 3 XP_016857129.1
XM_017001641.1 4944 UTR 3 XP_016857130.1
XM_017001642.1 4944 UTR 3 XP_016857131.1
Gene
ZYG11A
Gene Name
zyg-11 family member A, cell cycle regulator
There are no transcripts associated with this gene.

View Full Product Details